EMAIL THIS PAGE TO A FRIEND
The AAPS journal
The pharmacokinetics and interactions of ivermectin in humans--a mini-review.
PMID 18446504
Abstract
Ivermectin is an antiparasitic drug with a broad spectrum of activity, high efficacy as well as a wide margin of safety. Since 1987, this compound has a widespread use in veterinary medicine and it use has been extended in humans. Here we present a brief review of the information available regarding the pharmacokinetics and interactions of ivermectin in humans. Awareness of these characteristics could improve the clinical efficacy of Ivermectin. All Authors declare that they do not have any Conflict of interest and that the work is original. All Authors agree that the contents of the manuscript are confidential and will not be copyrighted, submitted, or published elsewhere (including the Internet), in any language, while acceptance by the Journal is under consideration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I8000010
Ivermectin, European Pharmacopoeia (EP) Reference Standard