EMAIL THIS PAGE TO A FRIEND
MMW Fortschritte der Medizin
[Recognizing simple clinical signs. When is an asthma attack an emergency?].
PMID 18447270
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4153
Prednisolone 21-hemisuccinate sodium salt, ≥90%, powder
C25H31NaO8