EMAIL THIS PAGE TO A FRIEND
Water research
Oxidation of chlorfenvinphos in ultrapure and natural waters by ozonation and photochemical processes.
PMID 18448145
Abstract
The chemical oxidation of the organophosphorus insecticide chlorfenvinphos, a priority pollutant in aquatic environments, has been conducted in ultrapure water, by means of single degradation agents (ozone and UV radiation), and by the Advanced Oxidation Processes constituted by combinations of these oxidants (O(3)/H(2)O(2) and UV/H(2)O(2)). The influence of the operating variables was discussed, and the degradation rates were evaluated by determining the rate constants for the reactions with ozone (k(O)(3)=3.7+/-0.2 L mol(-1)s(-1)) and OH radicals (k(OH)=(3.2+/-0.2)x10(9) L mol(-1)s(-1)), as well as the quantum yield for the photodegradation (around 0.1 mol E(-1), depending on the pH). Additionally, the ozonation of chlorfenvinphos in a natural water system (a surface water from a reservoir) was studied. The influence of the operating conditions on the insecticide removal efficiency was established, and the R(ct) parameter was evaluated. A kinetic model was proposed for the prediction of the elimination rate of chlorfenvinphos in the ozonation process and the results obtained reveal a good agreement between experimental results and predicted values.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36551
Chlorfenvinphos, PESTANAL®, analytical standard
C12H14Cl3O4P