EMAIL THIS PAGE TO A FRIEND
Pharmaceutical research
Flux across [corrected] microneedle-treated skin is increased by increasing charge of naltrexone and naltrexol in vitro.
PMID 18449628
Abstract
The purpose of this investigation was to evaluate the in vitro microneedle (MN) enhanced percutaneous absorption of naltrexone hydrochloride salt (NTX x HCl) compared to naltrexone base (NTX) in hairless guinea pig skin (GP) and human abdominal skin. In a second set of experiments, permeability of the major active metabolite 6-beta-naltrexol base (NTXOL) in the primarily unionized (unprotonated) form at pH 8.5 was compared to the ionized form (pH 4.5). In vitro fluxes of NTX, NTX.HCl and ionized and unionized NTXOL were measured through microneedle treated or intact full thickness human and GP skin using a flow through diffusion apparatus. Solubility and diffusion samples were analyzed by HPLC. Both GP and human skin show significant increases in flux when treated with 100 MN insertions as compared to intact full thickness skin when treated with NTX.HCl or ionized NTXOL (pH 4.5; p < 0.05). MN increased GP skin permeability for the hydrophilic HCL salt of NTX by tenfold and decreased lag time by tenfold too. Similar results were found using human skin, such that skin permeability to NTX.HCl was elevated to 7.0 x 10(-5) cm/h. Permeability of the primarily unionized (unprotonated) form of NTXOL at pH 8.5 was increased by MN only threefold and lag time was only modestly reduced. However, MN treatment with the primarily ionized (protonated) form of NTXOL at pH 4.5 increased skin permeability fivefold and decreased lag time fourfold. Enhancement was observed in vitro in both GP and human skin treated with MN compared to intact skin with the salt form of NTX and the ionized form of NTXOL. We conclude that transdermal flux can be optimized by using MN in combination with charged (protonated) drugs that have increased solubility in an aqueous patch reservoir and increased permeability through aqueous pathways created by MN in the skin.
Related Materials