EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Preparation, characterization and in vitro anticancer activity of platinum(II) complexes with N-Cyclohexyl-1,3-propanediamine as the carrier.
PMID 18451553
Abstract
New JM118 (active form of satraplatin) analogues with N-cyclohexyl-1,3-propanediamine (N-chpda) as the carrier, cis-[Pt(N-chpda)X2] (X2=2Cl(-) (1), oxalate (2), malonate (3), 1,1-cyclobutanedicarboxylate (CBDCA) (3), and 3-hydroxy-1,1-cyclobutanedicarboxylate(HO-CBDCA) (4)), have been synthesized and characterized by elemental analysis and spectroscopic data along with X-ray crystal structure for a representative compound cis-[Pt(N-chpda)Cl2]. The complexes have also been evaluated for their in vitro anticancer activity. All these analytical data are in good agreement with the structures of the desired compounds. The Pt(II) is in a square planar environment and is coordinated by a chelating N-chpda ligand and 2Cl(-) in cis position, and there are two crystallographically independent cis-[Pt(N-chpda)Cl2] molecules linked together by intermolecular N-H...Cl hydrogen bonds. Compounds 1 and 2 are very active against human lung cancer cell line (AGZY) and human lymphocytic leukemia cell line (Raji), and are much more active than carboplatin. Platinum(II) complexes with N-cyclohexyl-1,3-propanediamine is an alternative choice for mixed ammine/aminoplatinum anticancer drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C108057
N-Cyclohexyl-1,3-propanediamine, 99%
C9H20N2