EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
An efficient synthesis of indolo[3,2-a]carbazoles via the novel acid catalyzed reaction of indoles and diaryl-1,2-diones.
PMID 18452007
Abstract
A mild and efficient one pot method for the synthesis of indolo[3,2-a]carbazole derivatives by the para-toluenesulfonic acid catalyzed condensation of indole with acyclic 1,2-diones is described. With cyclobutene-1,2-diones the reaction afforded indole substituted carbazole derivatives in good yield.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5151
Benzil, 98%
C14H10O2