EMAIL THIS PAGE TO A FRIEND
Journal of environmental quality
Site-specific adsorption of 1,3-dinitrobenzene to bacterial surfaces: a mechanism of n-pi electron-donor-acceptor interactions.
PMID 18453403
Abstract
Surface and subsurface contamination with nitroaromatic compounds (NACs) has drawn considerable attention, and biosorption may play an important role in the fate and transport of these compounds in the environment. We studied the sorption of polar 1,3-dinitrobenzene (DNB) as a representative NAC and 2,6-dichlorobenzonitrile and nonpolar phenanthrene and 1,2,4,5-tetrachlorobenzene from the aqueous phase to two common bacteria, gram-negative Escherichia coli and gram-positive Bacillus subtilis. Sorption of DNB is highly nonlinear and is well described by the Langmuir model and shows the highest capacity among all tested solutes (up to 2.4% of E. coli biomass and 7.6% of B. subtilis biomass by weight) despite the lowest solute hydrophobicity. These results indicate that strong specific sorptive interactions exist between DNB and bacterial surfaces. We propose a mechanism of n-pi electron-donor-acceptor interactions between the oxygen electron pairs of deprotonated carboxyl groups (electron donors) of bacterial surfaces and DNB (electron acceptor). Biosorption of DNB increases with deprotonation of functional groups as pH increases, which rules out hydrophobic effects and H-bonding as major sorption driving forces because they are both favored by protonation of functional groups as pH decreases.