EMAIL THIS PAGE TO A FRIEND
Journal of midwifery & women's health
Traveler's diarrhea: an update on prevention and treatment.
PMID 18455100
Abstract
Traveler's diarrhea is a condition that disrupts work and vacation, and affects many women traveling from developed countries to developing countries. Specific food and hygiene education may help to decrease the occurrence of this common food and water-borne gastrointestinal infection. Probiotics rather than antibiotics are now among the recommendations for the prophylaxis of traveler's diarrhea. Treatment for traveler's diarrhea in mild cases may involve simply maintaining hydration. In moderate cases that disrupt the travel itinerary antibiotics and antimotility agents can be used.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
480789
Bismuth(III) subsalicylate, 99.9% trace metals basis
C7H5BiO4