EMAIL THIS PAGE TO A FRIEND
The international journal of neuropsychopharmacology
Enhanced alpha1 adrenergic sensitivity in sensorimotor gating deficits in neonatal ventral hippocampus-lesioned rats.
PMID 18460229
Abstract
Neonatal ventral hippocampus (nVH) lesion in rats is a widely used animal model of schizophrenia due to the predominantly post-pubertal emergence of many schizophrenia-like behaviours. Our previous studies have shown increased ligand binding of alpha1 adrenergic receptors (AR) in the frontal cortex of post-pubertal, but not pre-pubertal, nVH-lesioned rats, compared to sham-lesioned control rats. Moreover, pretreatment with the alpha1 adrenergic receptor antagonist prazosin reversed amphetamine-induced hyperlocomotion in controls, but failed to do so in lesioned animals. This led to our hypothesis that nVH lesions may lead to post-pubertal hyperactivity of alpha1 adrenergic receptors. In order to test the functional relevance of alpha1 adrenergic hyperactivity to schizophrenia-like behaviours of nVH-lesioned animals, we conducted prepulse inhibition (PPI) studies in post-pubertal (postnatal days 56-120) sham and lesioned animals in response to systemic injections of alpha1 adrenergic receptor antagonist and agonist, prazosin and cirazoline, respectively. Our results show that PPI deficits in nVH-lesioned animals were reversed with prazosin treatment, without a significant effect on PPI in sham animals. Further, at various doses, cirazoline had a significantly greater PPI disruptive effect in nVH-lesioned animals than in sham animals. Together, these results suggest that nVH-lesioned animals show a hyperactive alpha1 adrenergic receptor system that may mediate sensorimotor gating abnormalities reported in these animals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C223
Cirazoline hydrochloride, solid
C13H16N2O · HCl