EMAIL THIS PAGE TO A FRIEND
Molecular Biology of the Cell
The involvement of the tyrosine kinase c-Src in the regulation of reactive oxygen species generation mediated by NADPH oxidase-1.
PMID 18463161