EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A "jellyfish" shaped green emitting gallium(III)-containing metallomesogen.
PMID 18463757
Abstract
The synthesis of the first gallium(III)-based liquid crystal has been achieved grafting around the metal centre two chelating 2-methylquinolin-8-olate anions and one monodentate 3,4,5-tris(hexadecyloxy)benzoyloxy ligand, allowing the resulting complex to be a soft luminescent material with the typical high quantum yield of pentacordinated gallium species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Q2125
Quinaldine, analytical standard, ≥90% (GC)
C10H9N
22550
Quinaldine, ≥95.0% (GC)
C10H9N