EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Control of electronic structure of a six-coordinate iron(III) porphyrin radical by means of axial ligands.
PMID 18463758
Abstract
Addition of tert-butylisocyanide (tBuNC) to a CD2Cl2 solution of the bis(perchlorato)(meso-tetramesitylporphyrinato) iron(III) cation radical leads to the formation of the corresponding bis(adduct), [Fe(TMP)(tBuNC)2]2+, whose electronic structure is in sharp contrast to that of the corresponding imidazole(HIm) complex, [Fe(TMP)(HIm)2]2+; the former adopts the S = 0 while the latter exhibits the S = 1 electronic ground state.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
133280
Butyl isocyanide, 97%
C5H9N