EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Identification of the hepatocyte growth factor receptor as the c-met proto-oncogene product.
PMID 1846706
Abstract
Hepatocyte growth factor (HGF) is a plasminogen-like protein thought to be a humoral mediator of liver regeneration. A 145-kilodalton tyrosyl phosphoprotein observed in rapid response to HGF treatment of intact target cells was identified by immunoblot analysis as the beta subunit of the c-met proto-oncogene product, a membrane-spanning tyrosine kinase. Covalent cross-linking of 125I-labeled ligand to cellular proteins of appropriate size that were recognized by antibodies to c-met directly established the c-met product as the cell-surface receptor for HGF.