EMAIL THIS PAGE TO A FRIEND
Cell biology and toxicology
Neuropathological effect of carbamate molluscicides on the land snail, Eobania vermiculata.
PMID 18470629
Abstract
The present study was designed to investigate the neuropathological effect of the two carbamate pesticides: methomyl and methiocarb on the neurons of the buccal ganglia in the land snail Eobania vermiculata using topical application and baiting technique. Their in vivo effects on acetylcholinesterase (AChE, EC 3.1.1.7) activity were also investigated. Sublethal dose and concentration (1/4 LD(50) and 1/4 LC(50)) of both pesticides were used, and the experiment lasted for 14 days. Histopathological and ultrastuctural alterations in the buccal ganglia were more obvious after the baiting technique treatment than after the topical application method, and methomyl was found to be more toxic than methiocarb. These alterations included shrinkage of the perikarya of neurons, increased cytoplasmic basophilia, and extreme indentation of the plasma membrane. In addition, the nuclei appeared karyolitic, eccentric, and highly shrunken with an irregular nuclear envelope. The most outstanding symptom observed after topical application of methiocarb was a highly vacuolated cytoplasm with a peripheral increase in electron density associated with dense accumulations of free ribosomes. On the other hand, an increased number of lysosomes and autophagosomes were observed after topical application of methomyl. Mitochondrial damage, increased number of lipid droplets, and myelin figures were frequently observed in ganglia treated with either methomyl or methiocarb. Moreover, it was noticed that both compounds induced reductions in AChE activity. However, methomyl exhibited more potency in reducing AChE activity than methiocarb.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36152
Methiocarb, PESTANAL®, analytical standard
C11H15NO2S