EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of N-substituted indole-3-carboxylic acid derivatives via Cu(I)-catalyzed intramolecular amination of aryl bromides.
PMID 18471015
Abstract
A variety of N-alkylated and N-arylated derivatives of methyl 1 H-indole-3-carboxylate were synthesized efficiently via Ullmann-type intramolecular arylamination, using the CuI-K 3PO 4-DMF system. This catalytic amination procedure can be performed with good to high yields under mild conditions under an air atmosphere.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284734
Indole-3-carboxylic acid, ReagentPlus®, 99%
C9H7NO2