EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Simultaneous determination of piracetam and vincamine by spectrophotometric and high-performance liquid chromatographic methods.
PMID 18476342
Abstract
A mixture of piracetam and vincamine was determined by 3 different methods. The first was the determination of piracetam and vincamine using the ratio-spectra first-derivative (DD1) spectrophotometric technique at 209 and 293 nm in concentration ranges of 10-45 and 2-14 microg/mL with mean recoveries of 99.22 +/- 0.72 and 99.67 +/- 0.79%, respectively. The second method was based on the resolution of the 2 components by bivariate calibration depending on a mathematic algorithm that provides simplicity and rapidity. The method depended on quantitative evaluation of the absorbencies at 210 and 225 nm in concentration ranges of 5-45 and 2-14 microg/mL, with mean recoveries of 100.33 +/- 0.54 and 100.44 +/- 0.98% for piracetam and vincamine, respectively. The third method was reversed-phase liquid chromatography using 0.05 M potassium dihydrogen phosphate-methanol (50 + 50, v/v) as the mobile phase, with the pH adjusted to 3.5 with phosphoric acid. The eluent was monitored at 215 nm in concentration ranges of 5-100 and 2-200 microg/mL, with mean recoveries of 99.62 +/- 0.67 and 99.32 +/- 0.85% for piracetam and vincamine, respectively. The suggested procedures were checked using laboratory-prepared mixtures and were successfully applied for the analysis of their pharmaceutical preparation. The methods retained their accuracy and precision when applying the standard addition technique. The results obtained by applying the proposed methods were statistically analyzed and compared with those obtained by the manufacturer's method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
194719
Vincamine, 98%
C21H26N2O3