EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Stability of chosen beta-adrenolytic drugs of different polarity in basic environment.
PMID 18476343
Abstract
The stability of atenolol, acebutolol, and propranolol was investigated in sodium hydroxide solutions at concentrations of 0.1, 0.3, 0.5, and 1 M at 3 temperatures ranging from 37 to 95 degrees C. The degradation processes that occurred in drugs under investigation were described with kinetic parameters (k, t0.1, and t0.5) and energy of activation (Ea). It was found that the stability of the drugs increased toward lipophilic propranolol in the assumed experimental model. The rate constants k decreased, contrary to t0.1, t0.5, and Ea, which varied comparably to log P (partition coefficient), thus increasing from the most hydrophilic atenolol, through acebutolol of lower polarity, to the most lipophilic propranolol. The identification of degradation products was performed with the application of proton nuclear magnetic resonance spectrometry and thin-layer chromatography-densitometry and data from the literature.
Related Materials