EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Highly enantioselective synthesis of isoxazoline N-oxides.
PMID 18478708
Abstract
The reaction of cinchonidine (cinchonine)-derived ammonium salts with nitroolefins in the presence of Cs2CO3 to afford optically active isoxazoline N-oxides with excellent ee and high de values has been developed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27350
Cinchonidine, purum, ≥98.0% (dried material, NT)
C19H22N2O
C80407
Cinchonidine, 96%
C19H22N2O