EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Stereospecific deuteration of alpha-furanosyl azomycin nucleosides: a model reaction for tritium radiolabeling.
PMID 18479915
Abstract
Stereospecific synthesis of 1-alpha-d-(2-deuteroribofuranosyl)-2-nitroimidazole (2'-[(2)H]-alpha-AZR) is reported. This, deuteration was independent of the configuration of C-2' -OH group (arabinose or ribose) in sugar moiety of starting molecules. Slightly better yield (>37%) of the deuterated product, 6, from arabinosyl precursor in comparison to corresponding ribose precursor (29%) was obtained which may reflect better stereochemical availability of C-2' -OH in arabinose during oxidation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
195650
2-Nitroimidazole, 98%
C3H3N3O2