EMAIL THIS PAGE TO A FRIEND
Nihon yakurigaku zasshi. Folia pharmacologica Japonica
[Treatment of Alzheimer's disease: status quo and future considerations].
PMID 18480565
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B2753
Butyrylcholine chloride, ≥98%
C9H20ClNO2
20780
Butyrylcholine iodide, ≥99.0% (AT)
C9H20INO2