EMAIL THIS PAGE TO A FRIEND
Journal of Clinical Psychopharmacology
Effect of antipsychotics on peptides involved in energy balance in drug-naive psychotic patients after 1 year of treatment.
PMID 18480685