EMAIL THIS PAGE TO A FRIEND
Pest management science
A novel assay reveals the blockade of esterases by piperonyl butoxide.
PMID 18481337
Abstract
Conventional in vitro assays sometimes fail to reveal esterase inhibition by piperonyl butoxide (PBO), although synergism studies suggest loss of esterase-mediated sequestration of insecticide does take place. A new in vitro assay has been devised that routinely reveals binding between PBO and these esterases. The new 'esterase interference' assay detects the blockade of resistance-associated esterases in a species, Myzus persicae Sülzer, where this has not previously been seen. The assay also demonstrates directly the protective effect esterases may confer to target sites of insecticides. The new assay reveals esterase blockade by PBO and thus has the potential to be used as a high-throughput screening method for other potential synergists.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45331
Azamethiphos, PESTANAL®, analytical standard
C9H10ClN2O5PS