EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Preparation of new solid phase micro extraction fiber on the basis of atrazine-molecular imprinted polymer: application for GC and GC/MS screening of triazine herbicides in water, rice and onion.
PMID 18482598
Abstract
A simple polymerization strategy has been used to produce a monolithic solid phase micro extraction (SPME) fiber on the basis of molecularly imprinted polymer able to couple with GC and GC-MS for selective extraction and analysis of triazine herbicides. A fiber was produced by copolymerization of methacrylic acid-ethylene glycol dimethacrylate imprinted with atrazine. The effective factors influencing the polymerization have been investigated and are detailed here. At the optimum conditions the prepared fiber is firm, inexpensive, durable and thermally stable up to 280 degrees C which has vital importance in SPME coupled with GC or GC/MS. In addition, the influences of pH, extraction time and temperature on the extraction efficiency of analytes were optimized. Selectivity of prepared fibers in relation to triazine herbicides and some of the other pesticide has been investigated. The high extraction efficiency was obtained for atrazine, simazine, propazine, cyanazine, ametryn, terbutryn and prometryn yielding the detection limits of 20, 70, 80, 81, 69, 88 and 68 ng mL(-1), respectively and the high quantities of recoveries. The reliability of prepared fiber to extraction of atrazine and other analogues in real samples has been investigated and proved by implementation of SPME in spiked samples such as tap water, onion and rice.