Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment
SiOx layer as functional barrier in polyethylene terephthalate (PET) bottles against potential contaminants from post-consumer recycled PET.
PMID 18484307
The barrier effect of a silicon oxide (SiOx) coating on the inner surface of PET bottles, in terms of the ability to reduce the migration of post-consumer compounds from the PET bottle wall into food simulants (3% acetic acid and 10% ethanol), was investigated. The barrier effect was examined by artificially introducing model substances (surrogates) into the PET bottle wall to represent a worst-case scenario. Test bottles with three different spiking levels up to approximately 1000 mg kg(-1) per surrogate were blown and coated on the inner surface. The SiOx-coated bottles and the non-coated reference bottles were filled with food simulants. From the specific migration of the surrogates with different bottles wall concentrations, the maximum surrogate concentrations in the bottle wall corresponding to migration of 10 microg l(-1) were determined. It was shown that the SiOx coating layer is an efficient barrier to post-consumer compounds. The maximum bottle wall concentrations of the surrogates corresponding to migration of 10 microg l(-1) were in the range of 200 mg kg(-1) for toluene and approximately 900 mg kg(-1) for benzophenone. Consequently, the SiOx coating allows use of conventionally recycled post-consumer PET flakes (without a super-clean recycling process) for packaging aqueous and low alcoholic foodstuffs (under cold-fill conditions) and protects food from migration of unwanted contaminants from post-consumer PET.