EMAIL THIS PAGE TO A FRIEND
Tuberculosis (Edinburgh, Scotland)
Rifapentine.
PMID 18486059
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0533
Rifapentine
C47H64N4O12