EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and spectroscopic studies of biologically active compounds derived from oxalyldihydrazide and benzil, and their Cr(III), Fe(III) and Mn(III) complexes.
PMID 18486278
Abstract
A new series of complexes have been synthesized by template condensation of oxalyldihydrazide and benzil in methanolic medium in the presence of trivalent chromium, manganese and iron salts forming complexes of the type [M(C(32)H(24)N(8)O(4))X]X(2) where M = Cr(III), Mn(III), Fe(III) and X = Cl(-1), NO(3)(-1), CH(3)COO(-1). The complexes have been characterized with the help of elemental analyses, conductance measurements, magnetic susceptibility measurements, electronic, NMR, infrared and far infrared spectral studies. On the basis of these studies, a five coordinate square pyramidal geometry has been proposed for all these complexes. The biological activities of the metal complexes have been tested in vitro against a number of pathogenic bacteria to assess their inhibiting potential. Some of these complexes have been found to exhibit remarkable antibacterial activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5151
Benzil, 98%
C14H10O2
75740
Oxalyldihydrazide, purum, ≥98.0% (N)
C2H6N4O2
131296
Oxalyldihydrazide, 98%
C2H6N4O2