EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Levoglucosan and other cellulose markers in pyrolysates of Miocene lignites: geochemical and environmental implications.
PMID 18497150
Abstract
Using the pyrolysis-gas chromatography-mass spectrometry and off-line pyrolysis/silylation methods for lignites from three Miocene brown coal basins of Poland resulted in the characterization of many organic compounds, including dominant cellulose degradation products such as levoglucosan, 1,6-anhydro-beta-D-glucofuranose, and 1,4:3,6-dianhydroglucopyranose. Levoglucosan is a general source-specific tracer for wood smoke in the atmosphere and recent sediments. The presence of unusually high levels of this compound in brown coal pyrolysates suggests that a portion of this compound concentration in some airsheds may originate from lignite combustion. On the other hand, nonglucose anhydrosaccharides, in particular, mannosan and galactosan, typical of hemicellulose, are not detected in those lignite pyrolysates investigated. This indicates that mannosan and galactosan are better specific tracers for combustion of contemporary biomass in those regions were the utilization of brown coals containing fossilized cellulose is important.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
324620
Lithium bis(trimethylsilyl)amide, 97%
C6H18LiNSi2
577014
Lithium bis(trimethylsilyl)amide solution, 1 M in tert-butyl methyl ether
C6H18LiNSi2
224367
Lithium bis(trimethylsilyl)amide solution, 1.0 M in hexanes
C6H18LiNSi2
225770
Lithium bis(trimethylsilyl)amide solution, 1.0 M in THF
C6H18LiNSi2
577928
Lithium bis(trimethylsilyl)amide solution, 1 M in toluene
C6H18LiNSi2
771864
Lithium bis(trimethylsilyl)amide solution, 0.5 M in toluene
C6H18LiNSi2
766917
Lithium bis(trimethylsilyl)amide solution, 1.5 M in THF
C6H18LiNSi2
775053
Lithium bis(trimethylsilyl)amide solution, 0.5 M in 2-methyltetrahydrofuran
C6H18LiNSi2
324671
Potassium bis(trimethylsilyl)amide, 95%
C6H18KNSi2
277304
Potassium bis(trimethylsilyl)amide solution, 0.5 M in toluene
C6H18KNSi2
702722
Potassium bis(trimethylsilyl)amide solution, 1 M in THF
C6H18KNSi2
766941
Potassium bis(trimethylsilyl)amide solution, 1.0 M in methyl tert-butyl ether
C6H18KNSi2
775061
Potassium bis(trimethylsilyl)amide solution, 1.0 M in 2-methyltetrahydrofuran
C6H18KNSi2
235083
Sodium bis(trimethylsilyl)amide, 95%
C6H18NNaSi2
80631
Sodium bis(trimethylsilyl)amide solution, produced by Wacker Chemie AG, Burghausen, Germany, 40% in THF
C6H18NNaSi2
245585
Sodium bis(trimethylsilyl)amide solution, 1.0 M in THF
C6H18NNaSi2
368059
Sodium bis(trimethylsilyl)amide solution, 0.6 M in toluene
C6H18NNaSi2
771546
Sodium bis(trimethylsilyl)amide solution, 0.5 M in methyl tert-butyl ether
C6H18NNaSi2
547816
Tris[N,N-bis(trimethylsilyl)amide]europium(III), 98%
C18H54EuN3Si6
547824
Tris[N,N-Bis(trimethylsilyl)amide]gadolinium(III), 98%
C18H54GdN3Si6
547980
Tris[N,N-bis(trimethylsilyl)amide]samarium(III), 98%
C18H54N3Si6Sm
524514
Tris[N,N-bis(trimethylsilyl)amide]yttrium
C18H54N3Si6Y
702021
Tris[N,N-bis(trimethylsilyl)amide]yttrium, packaged for use in deposition systems
C18H54N3Si6Y