EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
The human cerebrospinal fluid metabolome.
PMID 18502700
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219835
2-Hydroxy-3-methylbutyric acid, 99%
C5H10O3
I9030
2-Propanol, meets USP testing specifications
C3H8O
650447
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
C3H8O
34863
2-Propanol, for HPLC, 99.9%
C3H8O
439207
2-Propanol, for HPLC, 99.5%
C3H8O
190764
2-Propanol, ACS reagent, ≥99.5%
C3H8O
673773
2-Propanol, ACS reagent, ≥99.5%
C3H8O
59300
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
C3H8O
33539
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
C3H8O
24137
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
C3H8O
109827
2-Propanol, Laboratory Reagent, ≥99.5%
C3H8O
166898
3-Hydroxybutyric acid, 95%
C4H8O3
K1750
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
C5H6O5
K1128
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
C5H6O5
75890
α-Ketoglutaric acid, ≥99.0% (T)
C5H6O5
695092
Acetic acid, ACS reagent, ≥99.7%
C2H4O2
A6283
Acetic acid, ReagentPlus®, ≥99%
C2H4O2
338826
Acetic acid, ≥99.99% trace metals basis
C2H4O2
33209
Acetic acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
C2H4O2
27225
Acetic acid, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
C2H4O2
27221
Acetic acid, puriss., 99-100%
C2H4O2
45754
Acetic acid solution, for HPLC
C2H4O2
650501
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
C3H6O
270725
Acetone, for HPLC, ≥99.9%
C3H6O
34850
Acetone, for HPLC, ≥99.8%
C3H6O
179124
Acetone, ACS reagent, ≥99.5%
C3H6O
320110
Acetone, ACS reagent, ≥99.5%
C3H6O
673781
Acetone, ACS reagent, ≥99.5%
C3H6O
534064
Acetone, histological grade, ≥99.5%
C3H6O
32201
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
C3H6O