EMAIL THIS PAGE TO A FRIEND
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan
[Investigation of ochratoxin a, B and citrinin contamination in various commercial foods].
PMID 18503248
Abstract
Ochratoxin A (OTA), ochratoxin B (OTB) and citrinin (CIT) in commercial foods were simultaneously determined and confirmed with high-performance liquid chromatography (HPLC) and liquid chromatography coupled with tandem mass spectrometry (LC/MS/MS). The samples examined were made up of cereal, fruit, coffee, and cacao products. The limits of quantification (S/N> or =10) of OTA, OTB and CIT were 0.1 microg/kg or less. Aflatoxins (AF), deoxynivalenol (DON) and fumonisins were also surveyed. Of 157 samples examined, 44 were contaminated with OTA at levels of 0.11 to 4.0 microg/kg. At least 2 positive samples were labeled as domestics. In most positive samples, the OTA level was low, less than 1 microg/kg. The highest incidence of OTA was observed in cacao powder (10/12), followed by instant coffee (5/7), cocoa (5/8) and raisin (6/13). OTB was found in fruit and cacao products containing relatively high levels of OTA. Co-occurrence of OTA, CIT and DON was found in cereal products, and co-occurrence of OTA and AF was found in cacao products. Approximately 30% of naturally contaminated OTA in roasted coffee bean moved into the extract solution when brewed with paper filter.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32411
Ochratoxin B solution, 10 μg/mL in acetonitrile, analytical standard
C20H19NO6