EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Adsorption of essential micronutrients by carbon nanotubes and the implications for nanotoxicity testing.
PMID 18504717
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
251917
Sulfanilic acid, ACS reagent, 99%
C6H7NO3S
86090
Sulfanilic acid, puriss. p.a., ≥99.0% (T)
C6H7NO3S