EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Total synthesis of (-)-corilagin.
PMID 18505255
Abstract
The synthesis of corilagin was achieved by the integration of the development of the oxidative coupling of the symmetrically protected gallates and the temporarily ring-opened synthetic route for the 3,6-hexahydroxydiphenoyl (HHDP) bridge. This is the first total synthesis of the 1C4/B-ellagitannins, which contain ring-flipped glucose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
75251
Corilagin, analytical standard
C27H22O18
G0424
Corilagin
C27H22O18