EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Fast stereoselective reactions in electrosprayed Co(II)/neurotransmitter nanodroplets.
PMID 18506238
Abstract
Collision induced dissociation (CID) of the m/z 479 ion, formed by ESI of Co(NO3)2-CH3OH solutions with either pure (1S,2S)-(+)-N-methylpseudoephedrine or its mixtures with (1S,2R)-(+)- or (1R,2S)-(-)-ephedrine, provides compelling evidence for fast, stereoselective reactions in Co(II)/neurotrasmitter(s) aggregates during solvent evaporation of the ESI droplets.