EMAIL THIS PAGE TO A FRIEND
Journal of Molecular Biology
Recruitment of CREB1 and histone deacetylase 2 (HDAC2) to the mouse Ltbp-1 promoter regulates its constitutive expression in a dioxin receptor-dependent manner.
PMID 18508077