EMAIL THIS PAGE TO A FRIEND
The Journal of Neuroscience
Toll-like receptor 2 acts as a natural innate immune receptor to clear amyloid beta 1-42 and delay the cognitive decline in a mouse model of Alzheimer's disease.
PMID 18509040