EMAIL THIS PAGE TO A FRIEND
Biology of reproduction
Molecular identification of lyso-glycerophosphocholines as endogenous immunosuppressives in bovine and rat gonadal fluids.
PMID 18509166
Abstract
The ability of the gametes to escape detection by the immune system is vital to successful human reproduction. Furthermore, the observed capacity of the testis in some species to support tissue grafts without rejection (immunological privilege) indicates that spermatogenic cells are protected by local immunoregulatory mechanisms. One of these mechanisms involves targeting T cells for inactivation and destruction within the testicular environment. Although the fluids of the testis and ovary surrounding the developing gametes contain soluble factors that inhibit T cells, the identity of the molecule(s) responsible for this activity has been unknown. Using a specific T-cell proliferation assay to monitor bioactivity, these molecules were purified from bovine ovarian follicular fluid by methanol extraction and sequential reverse-phase HPLC (RP-HPLC). All purified active fractions coincided with the elution position on RP-HPLC of several small molecules ranging in size from 496 to 522 Da. The same molecules were localized to the immunosuppressive fractions of rat testicular interstitial fluid. The active molecules were identified, using capillary electrophoresis electrospray ionization mass spectroscopy, as lyso-glycerophosphocholines (lyso-GPCs), namely, 1-palmitoyl-sn-glycero-3-phosphocholine, 1-oleoyl-sn-glycero-3-phosphocholine, a 18:2a/lyso-GPC (putatively, 1-linoleoyl-sn-glycero-3-phosphocholine), and a 20:4a/lyso-GPC (putatively, 1-arachidonyl-sn-glycero-3-phosphocholine). Comparison of the bioactivity and mass spectroscopy profiles of two of the purified molecules with their synthetic standards confirmed the identification. These molecules inhibit T-cell proliferation in response to activation and induce apoptosis of these cells in a time- and dose-dependent manner. The emergence of gonadal lyso-GPCs as potential regulators of critical immune events opens up new avenues of inquiry into the origins of autoimmune infertility and more generally into mechanisms of peripheral immunoregulation and the development of novel immunosuppressives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L1881
1-Oleoyl-sn-glycero-3-phosphocholine, synthetic, ≥99%
C26H52NO7P