EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Quercetin and rutin as potential sunscreen agents: determination of efficacy by an in vitro method.
PMID 18512988
Abstract
Given that flavonoids are known for their ultraviolet (UV)B photoprotective properties in plants that contain them, we chose to study quercetin (1) and rutin (2) as agents that could potentially be used in sunscreen products. These two substances proved to behave in similar ways. When incorporated in oil-in-water emulsions, at a concentration of 10% (w/w), 1 and 2 give sun protection factor (SPF) values similar to that of homosalate, a standard substance. These two flavonoids also provided a non-negligible level of photoprotection in the UVA range. When used in association with titanium dioxide, the SPF obtained was around 30.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1085
Homosalate, Pharmaceutical Secondary Standard; Certified Reference Material
C16H22O3