EMAIL THIS PAGE TO A FRIEND
The FEBS journal
Mammalian xanthine oxidoreductase - mechanism of transition from xanthine dehydrogenase to xanthine oxidase.
PMID 18513323
Abstract
Reactive oxygen species are generated by various biological systems, including NADPH oxidases, xanthine oxidoreductase, and mitochondrial respiratory enzymes, and contribute to many physiological and pathological phenomena. Mammalian xanthine dehydrogenase (XDH) can be converted to xanthine oxidase (XO), which produces both superoxide anion and hydrogen peroxide. Recent X-ray crystallographic and site-directed mutagenesis studies have revealed a highly sophisticated mechanism of conversion from XDH to XO, suggesting that the conversion is not a simple artefact, but rather has a function in mammalian organisms. Furthermore, this transition seems to involve a thermodynamic equilibrium between XDH and XO; disulfide bond formation or proteolysis can then lock the enzyme in the XO form. In this review, we focus on recent advances in our understanding of the mechanism of conversion from XDH to XO.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X2252
Xanthine Oxidase microbial, lyophilized powder, ≥7 units/mg solid