EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and biological activities of reveromycin A and spirofungin A derivatives.
PMID 18519164
Abstract
Various derivatives of reveromycin A, an inhibitor of eukaryotic cell growth, and spirofungin A, focusing on the 5S hydroxyl group and C18 hemisuccinyl group, were synthesized and their inhibitory effects on both the isoleucyl-tRNA synthetase activity and the survival of osteoclasts, and activities on the morphological reversion of src(ts)-NRK cells were examined. It was found that 2,3-dihydroreveromycin A is the promising derivative of reveromycin A based on the activity and stability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0654
Reveromycin A, ≥98% (HPLC), solid
C36H52O11