EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Spread of OXA-48-positive carbapenem-resistant Klebsiella pneumoniae isolates in Istanbul, Turkey.
PMID 18519712
Abstract
The first outbreak of carbapenem-resistant Klebsiella pneumoniae isolates producing the plasmid-encoded carbapenem-hydrolyzing oxacillinase OXA-48 is reported. The 39 isolates belonged to two different clones and were collected at the University Hospital of Istanbul, Turkey, from May 2006 to February 2007, and they coproduced various beta-lactamases (SHV-12, OXA-9, and TEM-1 for clone A and CTX-M-15, TEM-1, and OXA-1 for clone B).