EMAIL THIS PAGE TO A FRIEND
Journal of psychiatric practice
How multiple medication use evolves and the importance of therapeutic trials: the slippery slide.
PMID 18520787
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5536
Nefazodone hydrochloride, ≥98% (HPLC), solid
C25H32ClN5O2 · HCl