EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Incorporation of mitochondrial membrane proteins into liposomes containing acidic phospholipids.
PMID 185220
Abstract
Cytochrome oxidase, QH2-cytochrome c reductase, and the oligomycin-sensitive adenosine triphosphatase were incorporated into liposomes by a new procedure which yielded unidirectional orientation of the proteins. Cytochrome oxidase was reconstituted in the mitochrondrial orientation and the adenosine triphosphatase in the submitochondrial orientation. Reconstitutions were achieved by incubating the proteins at room temperature with liposomes which contained phosphatidylcholine, phosphatidylethanolamine, and an acidic phospholipid (cardiolipin, phosphatidylinositol, or phosphatidylserine). The incorporation occurred without added detergent or sonication. This incorporation procedure may serve as a model for the insertion of proteins in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63064
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Cl2Mg · 6H2O