EMAIL THIS PAGE TO A FRIEND
Vestnik khirurgii imeni I. I. Grekova
[Certain aspects of pathogenesis of hemorheological and trophic disturbances and their pharmacological correction in patients with chronic venous insufficiency].
PMID 18522184
Abstract
The work presents an analysis of results of conservative treatment of 48 patients aged from 32 through 76 years having postthrombophlebitic syndrome with symptoms of chronic venous insufficiency. The system of complex pharmacotherapy included phlebothropic medicines of flavonoid series (detralex) and of synthetic origin (tribenosid). The course of treatment was 2-3 months. Long-term results of the treatment were followed-up during the period from 6 months to 2 years in 44 patients. The complex treatment resulted in reliably less degree of clinical manifestations (chronic venous insufficiency). Recurrence of trophic ulcers appeared in 6 patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000165 Tribenoside, European Pharmacopoeia (EP) Reference Standard
C29H34O6