EMAIL THIS PAGE TO A FRIEND
Nutrition reviews
Update on alpha-linolenic acid.
PMID 18522621
Abstract
Consumption of omega 3 fatty acids is known to have health benefits. For many years, the importance of the only member of the omega 3 family considered to be essential, alpha-linolenic acid (ALA), has been overlooked. Current research indicates that ALA, along with its longer chain metabolites, may play an important role in many physiological functions. Potential benefits of ALA include cardioprotective effects, modulation of the inflammatory response, and a positive impact on both central nervous system function and behavior. Recommended levels for ALA intake have been set, yet the possible advantages of its consumption are just being revealed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62160
Linolenic acid, analytical standard
C18H30O2
62170
Linolenic acid, ~70% (GC)
C18H30O2