EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
DNA molecules on GaP (100) surfaces: spectroscopic characterization and biospecificity assessment.
PMID 18523943