EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
Antifeedant activity of some polygodial derivatives.
PMID 18533465
Abstract
Polygodial (1) and its derivatives acetal 2 (propylene) and 3 (ethylene) were prepared and their antifeedant activity and toxic effects evaluated on several insect species with different feeding ecologies (Spodoptera littoralis, Leptinotarsa decemlineata, Myzus persicae and Rhopalosiphum padi) along with that of polygonone (4). We also tested their selective cytotoxic effects on insect-derived (Spodoptera frugiperda ovarian Sf9 cells) and mammalian Chinese hamster ovary (CHO) cells. The antifeedant activity of these compounds was consistent with the proposed mode of action for antifeedant drimanes, i. e. adduct formation with amino groups for M. persicae and R. padi (dialdehyde > ketoaldehyde > aldehydeacetal). This was not the case for L. decemlineata, and the cytotoxic effects on insect-derived Sf9 and mammalian CHO cells (aldehydeacetal > dialdehyde > ketoaldehyde).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0049 Polygodial, ≥97% (HPLC)
C15H22O2