EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[New neolignan from seed of Myristica fragrans].
PMID 18533495
Abstract
To study the chemical constituents in the nutmeg (seed of Myristica fragrans). The chemical constituents were isolated by various column chromatographic methods and structurally elucidated by IR, NMR and MS evidences. Fifteen compounds were obtained and identified as myristicin (1), methyleugenol (2), safrole (3), 2, 3-dihydro-7-methoxy-2(3, 4-methylenedioxyphenyl)-3-methyl-5-(E) -propenyl-benzofuran (4), dehydrodiisoeugenol (5), 2, 3-dihydro-7-methoxy-2-(3-methoxy-4, 5-methylenedioxyphenyl) -3-methyl-5-(E)-propenyl-benzofuran (6), erythro-2-(4-allyl-2, 6-dimethoxyphenoxy)-1-(3, 4-dimetho- xyphenyl) propane (7), erythro-2-(4-allyl-2, 6-dimethoxyphenoxy)-1-(3, 4, 5-trimethoxyphenyl) propane (8), erythro-2-(4-allyl-2, 6-dimethoxyphenoxy)-1-(3, 4-dimethoxyphenyl) propan-1-ol acetate (9), erythro-2-(4-allyl-2, 6-dimethoxyphenoxy)-1-(3, 4-dimethoxyphenyl) propan-1-ol (10), erythro-2-(4-allyl-2, 6-dimethoxyphenoxy)-1-(3, 4, 5-trimethoxyphenyl) propan-1-ol (11), 5-methoxy-dehydrodiisoeugenol (12), erythro-2-(4-allyl-2, 6-dimethoxyphenoxy)-1-(4-hydroxy-3-methoxyphenyl)-propan-1-ol (13), guaiacin (14) and threo-2-(4-allyl-2, 6-dimethoxyphenoxy)-1-(3-methoxy-5-hydroxy-phenyl) propan-1-ol (15). Compound 15 is a new compound and named myrisisolignan. Compound 7 is isolated from the genus Myristica for the first time.
Related Materials