EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Ligand-to-metal ratio controlled assembly of nanoporous metal-organic frameworks.
PMID 18535699
Abstract
Two bilayered metal-organic frameworks with nanoporous channels were synthesized at different ligand-to-metal ratios, which demonstrated an interesting crystal-to-crystal transformation property and a special fluorescent response to the different guest molecules included.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45492
Fentin chloride, PESTANAL®, analytical standard
C18H15ClSn