EMAIL THIS PAGE TO A FRIEND
Journal of analytical toxicology
Error in the article: "driving under the influence of opiates: concentration relationships between morphine, codeine, 6-acetyl morphine, and ethyl morphine in blood".
PMID 18544226