EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Fate of the fungicide tolylfluanid in the pear cold stored in controlled or non controlled atmosphere.
PMID 1854993
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32060
Tolylfluanid, PESTANAL®, analytical standard
C10H13Cl2FN2O2S2