EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
The new NHS: restricting GPs' access to x ray services.
PMID 1855037
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219142
5-Bromopyrimidine, 97%
C4H3BrN2